Assay Details
| Gene Symbol: BOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558548.2 | XP_005558605.1 | 1 | 675 | 142 | |
| XM_005558549.2 | XP_005558606.1 | 1 | 675 | 142 | ||
| XM_015452250.1 | XP_015307736.1 | 1 | 675 | 142 | ||
| GenBank mRNA | AB171027.1 | - | 1 | 256 | 142 | |
More Information
Set Membership: |
|

BOD1
Genomic Map