Assay Details
| Gene Symbol: LOC102139374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015444355.1 | XP_015299841.1 | 3 | 617 | 89 | |
| Gene Symbol: CFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447396.1 | XP_015302882.1 | 11 | 2160 | 89 | |
More Information
Set Membership: |
|

LOC102139374
Genomic Map