Assay Details
Gene Symbol: LOC102126536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555085.2 | XP_005555142.1 | 3 | 537 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
