Assay Details
| Gene Symbol: LOC102139287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436652.1 | XP_015292138.1 | 4 | 2117 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102139287| Gene Symbol: LOC102139287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436652.1 | XP_015292138.1 | 4 | 2117 | 92 | |
Set Membership: |
|