Assay Details
| Gene Symbol: LOC102121713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565317.2 | XP_005565374.1 | 6 | 650 | 70 | |
| XM_005565319.2 | XP_005565376.1 | 6 | 738 | 70 | ||
| XM_005565320.2 | XP_005565377.1 | 6 | 588 | 70 | ||
| XM_005565321.2 | XP_005565378.1 | 5 | 545 | 70 | ||
| XM_005565322.2 | XP_005565379.1 | 5 | 642 | 70 | ||
More Information
Set Membership: |
|

LOC102121713
Genomic Map