Assay Details
| Gene Symbol: LOC102146804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433209.1 | XP_015288695.1 | 1 | 518 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015433209.1 transcripts.
Set Membership: |
|

LOC102146804
Genomic Map