Assay Details
| Gene Symbol: LOC102130187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441409.1 | XP_015296895.1 | 5 | 1118 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102130187| Gene Symbol: LOC102130187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441409.1 | XP_015296895.1 | 5 | 1118 | 80 | |
Set Membership: |
|