Assay Details
| Gene Symbol: TNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287736.1 | NP_001274665.1 | 1 - 2 | 476 | 70 | |
| GenBank mRNA | AB064994.1 | - | 911 | 70 | ||
| AB169384.1 | - | 1 - 2 | 476 | 70 | ||
More Information
Set Membership: |
|

TNP2
Genomic Map