Assay Details
| Gene Symbol: CYP4F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283131.1 | NP_001270060.1 | 10 - 11 | 1308 | 93 | |
| XM_015440164.1 | XP_015295650.1 | 10 - 11 | 1421 | 93 | ||
| GenBank mRNA | DQ074799.1 | - | 10 - 11 | 1308 | 93 | |
More Information
Set Membership: |
|

CYP4F3
Genomic Map