Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441011.1 | XP_015296497.1 | 4 | 2854 | 103 | |
| XM_015441012.1 | XP_015296498.1 | 5 | 2889 | 103 | ||
| XM_015441013.1 | XP_015296499.1 | 5 | 2859 | 103 | ||
| XM_015441014.1 | XP_015296500.1 | 4 | 2817 | 103 | ||
| XM_015441016.1 | XP_015296502.1 | 4 | 2805 | 103 | ||
| XM_015441017.1 | XP_015296503.1 | 5 | 2858 | 103 | ||
| XM_015441018.1 | XP_015296504.1 | 4 | 2646 | 103 | ||
| GenBank mRNA | AB174723.1 | - | 2 | 1916 | 103 | |
More Information
Set Membership: |
|

ZNF135
Genomic Map