Assay Details
| Gene Symbol: BTN3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319372.1 | NP_001306301.1 | 2 - 3 | 246 | 67 | |
| XM_015449782.1 | XP_015305268.1 | 2 - 3 | 239 | 67 | ||
| XM_015449783.1 | XP_015305269.1 | 2 - 3 | 239 | 67 | ||
| XM_015449785.1 | XP_015305271.1 | 2 - 3 | 239 | 67 | ||
| GenBank mRNA | AB060220.1 | - | 2 - 3 | 246 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001319372.1, XM_015449782.1, XM_015449783.1, XM_015449785.1 transcripts.
Set Membership: |
|

BTN3A2
Genomic Map