Assay Details
| Gene Symbol: SUOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319604.1 | NP_001306533.1 | 3 - 4 | 281 | 66 | |
| XM_005571162.2 | XP_005571219.1 | 4 - 5 | 450 | 66 | ||
| XM_005571163.2 | XP_005571220.1 | 4 - 5 | 511 | 66 | ||
| XM_005571164.2 | XP_005571221.1 | 3 - 4 | 287 | 66 | ||
| XM_005571166.2 | XP_005571223.1 | 2 - 3 | 220 | 66 | ||
| XM_015431015.1 | XP_015286501.1 | 2 - 3 | 208 | 66 | ||
| GenBank mRNA | AB125197.1 | - | 3 - 4 | 281 | 66 | |
More Information
Set Membership: |
|

SUOX
Genomic Map