Assay Details
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284782.1 | NP_001271711.1 | 1 - 2 | 259 | 99 | |
XM_005583345.2 | XP_005583402.1 | 2 - 3 | 135 | 99 | ||
GenBank mRNA | AB174160.1 | - | 1 - 2 | 259 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001284782.1, XM_005583345.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
