Assay Details
| Gene Symbol: NCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283548.1 | NP_001270477.1 | 274 | 106 | ||
| GenBank mRNA | AB168220.1 | - | 272 | 106 | ||
| AB169897.1 | - | 274 | 106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCL| Gene Symbol: NCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283548.1 | NP_001270477.1 | 274 | 106 | ||
| GenBank mRNA | AB168220.1 | - | 272 | 106 | ||
| AB169897.1 | - | 274 | 106 | |||