Assay Details
| Gene Symbol: RPLP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283165.1 | NP_001270094.1 | 1 - 2 | 100 | 127 | |
| XM_005576799.2 | XP_005576856.1 | 1 - 2 | 116 | 127 | ||
| GenBank mRNA | AB170183.1 | - | 1 - 2 | 100 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001283165.1, XM_005576799.2 transcripts.
Set Membership: |
|

RPLP2
Genomic Map