Assay Details
| Gene Symbol: B3GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283626.1 | NP_001270555.1 | 2 | 1118 | 108 | |
| GenBank mRNA | AB171124.1 | - | 2 | 1118 | 108 | |
More Information
Set Membership: |
|

B3GALT4
Genomic Map