Assay Details
| Gene Symbol: ARF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283692.1 | NP_001270621.1 | 5 | 915 | 76 | |
| XM_005541054.2 | XP_005541111.1 | 5 | 1295 | 76 | ||
| XM_005541056.2 | XP_005541113.1 | 5 | 1058 | 76 | ||
| GenBank mRNA | AB169501.1 | - | 5 | 1026 | 76 | |
| AB170156.1 | - | 5 | 917 | 76 | ||
| AB170250.1 | - | 5 | 915 | 76 | ||
More Information
Set Membership: |
|

ARF1
Genomic Map