Assay Details
| Gene Symbol: SMIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584956.2 | XP_005585013.1 | 2 - 3 | 297 | 61 | |
| XM_015438852.1 | XP_015294338.1 | 1 - 2 | 1209 | 61 | ||
More Information
Set Membership: |
|

SMIM5
Genomic Map