Assay Details
| Gene Symbol: LOC101866053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB173575.1 | - | 1 - 2 | 1058 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101866053| Gene Symbol: LOC101866053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB173575.1 | - | 1 - 2 | 1058 | 97 | |