Assay Details
| Gene Symbol: S1PR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542570.2 | XP_005542627.1 | 2 | 1377 | 55 | |
| XM_015432061.1 | XP_015287547.1 | 2 | 1247 | 55 | ||
| GenBank mRNA | AB173810.1 | - | 1 | 625 | 55 | |
More Information
Set Membership: |
|

S1PR1
Genomic Map