Assay Details
| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB179245.1 | - | 4 - 5 | 500 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WBP2NL| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB179245.1 | - | 4 - 5 | 500 | 75 | |
Set Membership: |
|