Assay Details
Gene Symbol: RBMXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283479.1 | NP_001270408.1 | 2 | 1121 | 95 | |
XM_015434095.1 | XP_015289581.1 | 2 | 1337 | 95 | ||
GenBank mRNA | AB168648.1 | - | 2 | 1121 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
