Assay Details
| Gene Symbol: RIOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561694.2 | XP_005561751.1 | 2 - 3 | 2801 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIOX1| Gene Symbol: RIOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561694.2 | XP_005561751.1 | 2 - 3 | 2801 | 54 | |
Set Membership: |
|