Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567591.2 | XP_005567648.2 | 2 - 3 | 157 | 109 | |
| XM_005567592.2 | XP_005567649.1 | 2 - 3 | 1792 | 109 | ||
| XM_005567595.2 | XP_005567652.1 | 2 - 3 | 264 | 109 | ||
| XM_015457511.1 | XP_015312997.1 | 2 - 3 | 1940 | 109 | ||
More Information
Set Membership: |
|

GAL3ST1
Genomic Map