Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589851.2 | XP_005589908.1 | 2 - 3 | 587 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005589851.2 transcripts.
Set Membership: |
|

NTF4
Genomic Map