Assay Details
| Gene Symbol: LOC107126439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430904.1 | XP_015286390.1 | 2 - 3 | 303 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015430904.1 transcripts.
Set Membership: |
|

LOC107126439
Genomic Map