Assay Details
| Gene Symbol: CREB3L2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448099.1 | XP_015303585.1 | 1 - 2 | 51 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CREB3L2-AS1| Gene Symbol: CREB3L2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448099.1 | XP_015303585.1 | 1 - 2 | 51 | 68 | |
Set Membership: |
|