Assay Details
| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430081.1 | XP_015285567.1 | 1 | 620 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTSF1L| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430081.1 | XP_015285567.1 | 1 | 620 | 89 | |
Set Membership: |
|