Assay Details
| Gene Symbol: LOC102129056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570543.2 | XP_005570600.1 | 2 - 3 | 505 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102129056| Gene Symbol: LOC102129056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570543.2 | XP_005570600.1 | 2 - 3 | 505 | 107 | |
Set Membership: |
|