Assay Details
| Gene Symbol: PAQR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546006.2 | XP_005546063.2 | 2 - 3 | 1503 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAQR9| Gene Symbol: PAQR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546006.2 | XP_005546063.2 | 2 - 3 | 1503 | 95 | |
Set Membership: |
|