Assay Details
| Gene Symbol: CRYGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433141.1 | XP_015288627.1 | 1 - 2 | 265 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRYGA| Gene Symbol: CRYGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015433141.1 | XP_015288627.1 | 1 - 2 | 265 | 82 | |