Assay Details
| Gene Symbol: IGLL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456984.1 | XP_015312470.1 | 1 - 2 | 114 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGLL5| Gene Symbol: IGLL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456984.1 | XP_015312470.1 | 1 - 2 | 114 | 77 | |
Set Membership: |
|