Assay Details
Gene Symbol: KHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449184.1 | XP_015304670.1 | 3 - 4 | 520 | 84 | |
XM_015449185.1 | XP_015304671.1 | 3 - 4 | 693 | 84 | ||
XM_015449186.1 | XP_015304672.1 | 4 - 5 | 702 | 84 | ||
XM_015449187.1 | XP_015304673.1 | 4 - 5 | 402 | 84 | ||
XM_015449188.1 | XP_015304674.1 | 5 - 6 | 825 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
