Assay Details
| Gene Symbol: LOC102121175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452823.1 | XP_015308309.1 | 3 - 4 | 344 | 84 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102121175| Gene Symbol: LOC102121175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452823.1 | XP_015308309.1 | 3 - 4 | 344 | 84 | |