Assay Details
| Gene Symbol: EDDM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015453505.1 | XP_015308991.1 | 1 | 226 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDDM3A| Gene Symbol: EDDM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015453505.1 | XP_015308991.1 | 1 | 226 | 66 | |
Set Membership: |
|