Assay Details
| Gene Symbol: CHST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443194.1 | XP_015298680.1 | 1 | 721 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015443194.1 transcripts.
Set Membership: |
|

CHST5
Genomic Map