Assay Details
| Gene Symbol: LOC107127181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015434029.1 | XP_015289515.1 | 2 - 3 | 3180 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107127181| Gene Symbol: LOC107127181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015434029.1 | XP_015289515.1 | 2 - 3 | 3180 | 97 | |