Assay Details
| Gene Symbol: LOC102123204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454818.1 | XP_015310304.1 | 46 - 47 | 2647 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102123204| Gene Symbol: LOC102123204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454818.1 | XP_015310304.1 | 46 - 47 | 2647 | 58 | |
Set Membership: |
|