Assay Details
| Gene Symbol: TBC1D29P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015438031.1 | XP_015293517.1 | 8 - 9 | 709 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D29P| Gene Symbol: TBC1D29P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015438031.1 | XP_015293517.1 | 8 - 9 | 709 | 66 | |
Set Membership: |
|