Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441096.1 | XP_015296582.1 | 1 - 2 | 191 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441096.1 transcripts.
Set Membership: |
|

C1QA
Genomic Map