Assay Details
| Gene Symbol: LOC102126622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447075.1 | XP_015302561.1 | 1 - 2 | 755 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102126622| Gene Symbol: LOC102126622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447075.1 | XP_015302561.1 | 1 - 2 | 755 | 68 | |
Set Membership: |
|