Assay Details
| Gene Symbol: LOC102119644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015453039.1 | XP_015308525.1 | 2 - 3 | 157 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015453039.1 transcripts.
Set Membership: |
|

LOC102119644
Genomic Map