Assay Details
| Gene Symbol: AJM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436451.1 | XP_015291937.1 | 3 - 4 | 1705 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AJM1| Gene Symbol: AJM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436451.1 | XP_015291937.1 | 3 - 4 | 1705 | 98 | |
Set Membership: |
|