Assay Details
| Gene Symbol: LOC107129460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448914.1 | XP_015304400.1 | 3 - 4 | 1239 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107129460| Gene Symbol: LOC107129460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448914.1 | XP_015304400.1 | 3 - 4 | 1239 | 71 | |
Set Membership: |
|