Assay Details
| Gene Symbol: LOC102140589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456086.1 | XP_015311572.1 | 6 - 7 | 666 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102140589| Gene Symbol: LOC102140589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015456086.1 | XP_015311572.1 | 6 - 7 | 666 | 103 | |
Set Membership: |
|