Assay Details
| Gene Symbol: LOC102137288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437232.1 | XP_015292718.1 | 1 - 2 | 128 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015437232.1 transcripts.
Set Membership: |
|

LOC102137288
Genomic Map