Assay Details
| Gene Symbol: LOC102137288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437232.1 | XP_015292718.1 | 3 - 4 | 410 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102137288| Gene Symbol: LOC102137288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437232.1 | XP_015292718.1 | 3 - 4 | 410 | 64 | |
Set Membership: |
|