Assay Details
| Gene Symbol: LOC107129459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448909.1 | XP_015304395.1 | 1 - 2 | 496 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107129459| Gene Symbol: LOC107129459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448909.1 | XP_015304395.1 | 1 - 2 | 496 | 94 | |
Set Membership: |
|