Assay Details
| Gene Symbol: HUS1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554178.2 | XP_005554235.1 | 3 - 4 | 275 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005554178.2 transcripts.
Set Membership: |
|

HUS1B
Genomic Map