Assay Details
| Gene Symbol: LOC102123076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542560.2 | XP_005542617.1 | 8 - 9 | 1167 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102123076| Gene Symbol: LOC102123076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542560.2 | XP_005542617.1 | 8 - 9 | 1167 | 128 | |
Set Membership: |
|